missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-nitropyridine, 98%, Thermo Scientific™

Product Code. 10298423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10298423 5 g Glass bottle 5g
10221622 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10298423 Supplier Thermo Scientific Acros Supplier No. 360590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 15862-31-4
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.02
MDL Number MFCD00955628
InChI Key OFXNHXMPRZDIDM-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
PubChem CID 2734412
IUPAC Name 3-bromo-5-nitropyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 215°C to 219°C
Color Beige to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 218.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-3-bromo-5-nitropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.