missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-methylpyridine, 98%, Thermo Scientific™

Product Code. 10587953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10587953 1 g Glass bottle 1g
10092021 5 g Glass bottle 5g
10239543 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10587953 Supplier Thermo Scientific Acros Supplier No. 348590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17282-00-7
Molecular Weight (g/mol) 187.04
MDL Number MFCD00068231
InChI Key NDPKXEWDWTZBDG-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-methylpyridine, 2-amino-3-bromo-5-picoline, 6-amino-5-bromo-3-picoline, 3-bromo-5-methyl-2-pyridinamine, 2-amino-5-methyl-3-bromopyridine, 2-pyridinamine, 3-bromo-5-methyl, 3-bromo-5-methyl-pyridin-2-ylamine, 3-bromo-5-methyl-2-pyridylamine, 3-bromo-5-methylpyridin-2-ylamine, 3-bromo-5-methylpyridin-1-ium-2-amine
PubChem CID 519380
IUPAC Name 3-bromo-5-methylpyridin-2-amine
SMILES CC1=CC(=C(N=C1)N)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.0°C to 57.0°C
Color Beige to Yellow or Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H7BrN2
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 187.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-bromo-5-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.