missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,6-difluorobenzonitrile, 97%

Product Code. 15475648
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475648 250 mg 250mg
15465648 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475648 Supplier Thermo Scientific Alfa Aesar Supplier No. H59362.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 190011-81-5 | C7H4F2N2 | 154.12 g/mol

2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,6-difluorobenzonitrile is used in cosmetic and pharmaceutical intermediates

Solubility
Miscible with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190011-81-5
Molecular Formula C7H4F2N2
Molecular Weight (g/mol) 154.12
MDL Number MFCD13178244
InChI Key ZPQFHGOBMGGBEP-UHFFFAOYSA-N
Synonym 2-amino-3,6-difluoro-benzonitrile, 2-cyano-3,6-difluoroaniline, 2-amino-3,6-difluorobenzonitrile, benzonitrile, 2-amino-3,6-difluoro-9ci
PubChem CID 45090591
IUPAC Name 2-amino-3,6-difluorobenzonitrile
SMILES NC1=C(F)C=CC(F)=C1C#N

Specifications

Quantity 250 mg
UN Number UN3439
Solubility Information Miscible with water.
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Amino-3,6-difluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.