missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichlorobenzonitrile, 97%, Thermo Scientific™

Catalog Number 15421799
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
36764-94-0
C7H4Cl2N2
187.02
MFCD00158947
ZHKNDJRPOVUPMT-UHFFFAOYSA-N
2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile
737215
2-amino-3,5-dichlorobenzonitrile
NC1=C(Cl)C=C(Cl)C=C1C#N
This item is not returnable. View return policy
36764-94-0
C7H4Cl2N2
187.02
MFCD00158947
ZHKNDJRPOVUPMT-UHFFFAOYSA-N
2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile
737215
2-amino-3,5-dichlorobenzonitrile
NC1=C(Cl)C=C(Cl)C=C1C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 123°C to 127°C (literature)
Quantity 1 g
Beilstein 2720575
Formula Weight 187.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3,5-dichlorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'