missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichlorobenzoic acid, 97%

Product Code. 11428806
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428806 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428806 Supplier Thermo Scientific Alfa Aesar Supplier No. A16969.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2789-92-6 | C7H5Cl2NO2 | 206.02 g/mol

2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2789-92-6
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00017088
InChI Key KTHTXLUIEAIGCD-UHFFFAOYSA-N
Synonym 3,5-dichloroanthranilic acid, benzoic acid, 2-amino-3,5-dichloro, anthranilic acid, 3,5-dichloro, 3,5-dichloro-2-aminobenzoic acid, 2-amino-3,5-dichloro-benzoic acid, timtec-bb sbb003493, 2-amino-3,5-dichlorobenzoicacid, acmc-209gzl, rarechem al bo 0770, buttpark 49\07-46
PubChem CID 76036
IUPAC Name 2-amino-3,5-dichlorobenzoic acid
SMILES NC1=C(Cl)C=C(Cl)C=C1C(O)=O

Specifications

Melting Point 225°C to 227°C
Quantity 250 g
Beilstein 779100
Solubility Information Insoluble in water.
Formula Weight 206.03
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dichlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.