missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyridine, 97%

Product Code. 11420633
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420633 100 g 100g
11400633 5 g 5g
11410633 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420633 Supplier Thermo Scientific Alfa Aesar Supplier No. A12706.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35486-42-1 | C5H4Br2N2 | 251.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35486-42-1
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00038041
InChI Key WJMJWMSWJSACSN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyridine, 3,5-dibromo-2-pyridylamine, 2-pyridinamine, 3,5-dibromo, 3,5-dibromo-2-pyridinamine, 3,5-dibromo-pyridin-2-ylamine, 3,5-dibromo-2-pyridinylamine, 2-amino-3,5-dibromo pyridine, 3,5-dibrompyridin-2-amin, pubchem1130, maybridge1_001693
PubChem CID 98851
IUPAC Name 3,5-dibromopyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Br

Specifications

Melting Point 102°C to 105°C
Quantity 100 g
Beilstein 119390
Formula Weight 251.92
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dibromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.