missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromopyridin-2-amine, 97%, Thermo Scientific™

Product Code. 10565134
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10565134 25 g 25g
10555134 10 g 10g
10426784 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10565134 Supplier Thermo Scientific Maybridge Supplier No. BTB07310EB
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35486-42-1
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00038041
InChI Key WJMJWMSWJSACSN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyridine, 3,5-dibromo-2-pyridylamine, 2-pyridinamine, 3,5-dibromo, 3,5-dibromo-2-pyridinamine, 3,5-dibromo-pyridin-2-ylamine, 3,5-dibromo-2-pyridinylamine, 2-amino-3,5-dibromo pyridine, 3,5-dibrompyridin-2-amin, pubchem1130, maybridge1_001693
PubChem CID 98851
IUPAC Name 3,5-dibromopyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula Br2(C5H2N)NH2
Quantity 25 g
Formula Weight 251.91
Percent Purity 97%
Chemical Name or Material 3, 5-Dibromopyridin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.