missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromobenzoic acid, Thermo Scientific™

Product Code. p-2697694
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10512282

Brand: Thermo Scientific Maybridge BTB01924EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 609-85-8
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
MDL Number MFCD00017087
InChI Key WNABMWFLKQEGCP-UHFFFAOYSA-N
Synonym 3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
PubChem CID 219491
IUPAC Name 2-amino-3,5-dibromobenzoic acid
SMILES NC1=C(Br)C=C(Br)C=C1C(O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 294.93
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Amino-3, 5-dibromobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.