missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromobenzoic acid, Thermo Scientific™

Product Code. 10392305
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10392305 25 g 25g
10512282 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10392305 Supplier Thermo Scientific Maybridge Supplier No. BTB01924EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 609-85-8
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
MDL Number MFCD00017087
InChI Key WNABMWFLKQEGCP-UHFFFAOYSA-N
Synonym 3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
PubChem CID 219491
IUPAC Name 2-amino-3,5-dibromobenzoic acid
SMILES NC1=C(Br)C=C(Br)C=C1C(O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Quantity 25 g
Formula Weight 294.93
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Amino-3, 5-dibromobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.