missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromobenzaldehyde, Mikromol™

Product Code. 17423752
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17423752 100 mg 100mg
17922446 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17423752 Supplier Mikromol Supplier No. MM0005.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Amino-3,5-dibromobenzaldehyde
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-3,5-dibromobenzaldehyde
CAS 50910-55-9
Analyte or Component Names 2-Amino-3,5-dibromobenzaldehyde
Shipping Condition Room Temperature
Molecular Formula C7 H5 Br2 N O
Certifications/Compliance ISO 17025
Synonym Ambroxol Hydrochloride Imp. E (EP), Ambroxol Imp. E (EP), Bromhexine Hydrochloride Imp. B (EP), Bromhexine Imp. B (EP), 2-Amino-3,5-dibromobenzaldehyde, Ambroxol Hydrochloride Impurity E, Bromhexine Hydrochloride Impurity B, Ambroxol Impurity E, Bromhexine Impurity B
InChI Formula InChI=1S/C7H5Br2NO/c8-5-1-4(3-11)7(10)6(9)2-5/h1-3H,10H2
SMILES Nc1c(Br)cc(Br)cc1C=O
IUPAC Name 2-amino-3,5-dibromobenzaldehyde
Molecular Weight (g/mol) 278.93
Quantity 100 mg
Formula Weight 276.8738 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.