missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-(4-bromobenzoyl)benzoic Acid, TRC

Product Code. 30010432
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010432 50 mg 50mg
30009360 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010432 Supplier Toronto Research Chemicals Supplier No. TRCA59384050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 241496-82-2
Molecular Formula C14 H10 Br N O3
Molecular Weight (g/mol) 320.14
IUPAC Name 2-amino-3-(4-bromobenzoyl)benzoic acid
SMILES Nc1c(cccc1C(=O)c2ccc(Br)cc2)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H10BrNO3/c15-9-6-4-8(5-7-9)13(17)10-2-1-3-11(12(10)16)14(18)19/h1-7H,16H2,(H,18,19)
Formula Weight 318.9844
Chemical Name or Material 2-Amino-3-(4-bromobenzoyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.