missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline, TRC

Product Code. 30008616
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008616 25 mg 25mg
30008765 10 mg 10mg
30012506 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008616 Supplier Toronto Research Chemicals Supplier No. TRCA63100025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95896-78-9
Molecular Formula C12 H13 N5
Molecular Weight (g/mol) 227.27
Synonym 3H-Imidazo[4,5-f]quinoxalin-2-amine, 3,4,8-trimethyl-, 3,4,8-Trimethyl-3H-imidazo[4,5-f]quinoxalin-2-amine, 2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline, 2-Amino-3,4,8-trimethylimidazo[4,5-f]quinoxaline, 4,8-DiMeIQx
IUPAC Name 3,4,8-trimethylimidazo[4,5-f]quinoxalin-2-amine
SMILES Cc1cnc2cc(C)c3c(nc(N)n3C)c2n1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H13N5/c1-6-4-8-9(15-7(2)5-14-8)10-11(6)17(3)12(13)16-10/h4-5H,1-3H3,(H2,13,16)
Formula Weight 227.1171
Percent Purity >95%
Chemical Name or Material 2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.