missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,4,5-trimethoxybenzoic acid, 97%

Artikelnummer. 11457936
Ansicht ändern
Click to view available options
Quantity:
1 g
5 g
25 g
Packungsgröße:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
11457936 25 g 25g
11437936 1 g 1g
11447936 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457936 Supplier Thermo Scientific Alfa Aesar Supplier No. A14042.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61948-85-4 | C10H13NO5 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61948-85-4
Molecular Formula C10H13NO5
MDL Number MFCD00051630
InChI Key JSHSRQCOCMIIPA-UHFFFAOYSA-N
PubChem CID 96602
IUPAC Name 2-amino-3,4,5-trimethoxybenzoic acid

Specifications

Quantity 25 g
Beilstein 3353856
Synonym 3,4,5-trimethoxyanthranilic acid, benzoic acid,2-amino-3,4,5-trimethoxy, benzoic acid, 2-amino-3,4,5-trimethoxy, maybridge1_001555, acmc-1b683, 2-amino-3,4,5-trimethoxybenzoicacid, 2-amino-3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(C(O)=O)=C(N)C(OC)=C1OC
Molecular Weight (g/mol) 227.22
Formula Weight 227.22
Percent Purity 0.97
Chemical Name or Material 2-Amino-3,4,5-trimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.