missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-oxo-acetic Acid Ethyl Ester, TRC

Product Code. 30308805
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30308805 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308805 Supplier Toronto Research Chemicals Supplier No. TRCA61861010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-2-oxo-acetic Acid Ethyl Ester
CAS 617-36-7
Molecular Formula C4H7NO3
Quantity 10 g
Synonym Acetic acid, aminooxo-, ethyl ester (9CI), Oxamic acid, ethyl ester (6CI,7CI,8CI), 2-Amino-2-oxoacetic acid ethyl ester, Ethoxalamide, Ethyl 2-amino-2-oxoacetate, Ethyl oxamate, NSC 48381, O-Ethyl oxamate, Oxalic acid monoethyl ester amide, Oxamethane
InChI Formula InChI=1S/C4H7NO3/c1-2-8-4(7)3(5)6/h2H2,1H3,(H2,5,6)
SMILES CCOC(=O)C(=O)N
IUPAC Name ethyl 2-amino-2-oxoacetate
Molecular Weight (g/mol) 117.1
Formula Weight 117.0426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.