missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methylpropane Hydrochloride, TRC

Product Code. 30330691
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330691 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330691 Supplier Toronto Research Chemicals Supplier No. TRCA89817050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-2-methylpropane Hydrochloride
CAS 10017-37-5
Molecular Formula C4H11N.HCl
Quantity 50 mg
Synonym 2-Propanamine, 2-methyl-, hydrochloride (1:1), 2-Propanamine, 2-methyl-, hydrochloride (9CI), tert-Butylamine, hydrochloride (8CI), 1,1-Dimethylethylamine hydrochloride, 2-Methylpropan-2-amine hydrochloride, t-Butylamine hydrochloride, tert-Butylammonium chloride
InChI Formula InChI=1S/C4H11N.ClH/c1-4(2,3)5;/h5H2,1-3H3;1H
SMILES Cl.CC(C)(C)N
IUPAC Name 2-methylpropan-2-amine;hydrochloride
Molecular Weight (g/mol) 109.5978
Formula Weight 109.0658
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.