missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol, 99%

Product Code. 10247063 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10247063 2.5 kg Plastic Bottle 2.5kg
10704021 100 g Glass bottle 100g
10366400 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10247063 Supplier Thermo Scientific Acros Supplier No. 104050025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-69-5 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-69-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
MDL Number MFCD00004678
InChI Key UXFQFBNBSPQBJW-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
PubChem CID 1531
ChEBI CHEBI:991
IUPAC Name 2-amino-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 108.0°C to 110.0°C
Color White
Boiling Point 151.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula HOCH2C(CH3)(NH2)CH2OH
Quantity 2.5 kg
Beilstein 04, 303
Fieser 02,24
Merck Index 15, 445
Solubility Information Solubility in water: 2500g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 105.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-methyl-1, 3-propanediol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.