missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-ethyl-1,3-propanediol, 97%

Product Code. 11410328
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410328 500 g 500g
11400328 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410328 Supplier Thermo Scientific Alfa Aesar Supplier No. B24509.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-70-8 | C5H13NO2 | 119.164 g/mol

It is used in pharmaceutical industry and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry and in medicine.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

Density 1.099
Boiling Point 152°C to 153°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.488
Quantity 500 g
Sensitivity Hygroscopic
Merck Index 14,439
Solubility Information Fully miscible in water.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material 2-Amino-2-ethyl-1,3-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-2-ethyl-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-2-ethyl-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.