missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-ethyl-1,3-propanediol, 97%

Product Code. 11400328
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400328 100 g 100g
11410328 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400328 Supplier Thermo Scientific Alfa Aesar Supplier No. B24509.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-70-8 | C5H13NO2 | 119.164 g/mol

It is used in pharmaceutical industry and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry and in medicine.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

Density 1.099
Boiling Point 152°C to 153°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.488
Quantity 100 g
Sensitivity Hygroscopic
Merck Index 14,439
Solubility Information Fully miscible in water.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material 2-Amino-2-ethyl-1,3-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.