missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-cyanoacetamide, 97+%

Product Code. 10168152 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10168152

Brand: Thermo Scientific Acros 382760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6719-21-7 | C3H5N3O | 99.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6719-21-7
Molecular Formula C3H5N3O
Molecular Weight (g/mol) 99.09
MDL Number MFCD00040532
InChI Key JRWAUKYINYWSTA-UHFFFAOYSA-N
Synonym aminocyanoacetamide, 3-nitriloalaninamide, acetamide, 2-amino-2-cyano, 2-aminocyanoacetamide, aminocyanacetamide, 2-amino-cyanoacetamide, 2-amino-2cyanoacetamide, a-amino-a-cyanoacetamide, 2-amino-2-cyanacetamide, 2-amino 2-cyano acetamide
PubChem CID 98341
IUPAC Name 2-amino-2-cyanoacetamide
SMILES C(#N)C(C(=O)N)N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 120°C to 124°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula NCCH(NH2)CONH2
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether and methanol
Formula Weight 99.09
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 2-Amino-2-cyanoacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-2-cyanoacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.