missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2'-chloro-5-nitrobenzophenone, 98%

Product Code. 15442759
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442759 100 g 100g
15422759 5 g 5g
15432759 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442759 Supplier Thermo Scientific Alfa Aesar Supplier No. H66440.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2011-66-7 | C13H9ClN2O3 | 276.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2011-66-7
Molecular Formula C13H9ClN2O3
Molecular Weight (g/mol) 276.68
MDL Number MFCD00792453
InChI Key GRDGBWVSVMLKBV-UHFFFAOYSA-N
Synonym 2-amino-2'-chloro-5-nitrobenzophenone, 2-amino-5-nitrophenyl 2-chlorophenyl methanone, 2-amino-5-nitro-2'-chlorobenzophenone, 2-amino-2'-chloro-5-nitro benzophenone, 2-2-chlorobenzoyl-4-nitroaniline, methanone, 2-amino-5-nitrophenyl 2-chlorophenyl, 2-amino-5-nitrophenyl-2-chlorophenyl methanone, 2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone, 2-amino-5-nitro-2'-chloro benzophenone, pubchem3255
PubChem CID 74830
IUPAC Name (2-amino-5-nitrophenyl)-(2-chlorophenyl)methanone
SMILES NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 119°C to 123°C
Color Yellow
Quantity 100 g
Beilstein 2814216
Sensitivity Air sensitive
Formula Weight 276.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-2'-chloro-5-nitrobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.