missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-2-(2-chlorophenyl)acetic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-Amino-2-(2-chlorophenyl)acetic Acid
Specifications
Specifications
| Chemical Name or Material | 2-Amino-2-(2-chlorophenyl)acetic Acid |
| CAS | 88744-36-9 |
| Analyte or Component Names | 2-Amino-2-(2-chlorophenyl)acetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H8 Cl N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzeneacetic acid, α-amino-2-chloro-, Glycine, 2-(o-chlorophenyl)- (7CI), (R,S)-α-Amino-α-(2-chlorophenyl)acetic acid, 2-(2-Chlorophenyl)glycine, 2-Amino-2-(2-chlorophenyl)acetic acid, NSC 154921 |
| InChI Formula | InChI=1S/C8H8ClNO2/c9-6-4-2-1-3-5(6)7(10)8(11)12/h1-4,7H,10H2,(H,11,12) |
| SMILES | NC(C(=O)O)c1ccccc1Cl |
| IUPAC Name | 2-amino-2-(2-chlorophenyl)acetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction