missing translation for 'onlineSavingsMsg'
Learn More

(2-Amino-1H-benzimidazol-5-yl)phenylmethanone, Mikromol™

Product Code. 17913976
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913976 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913976 Supplier Mikromol Supplier No. MM0882.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2-Amino-1H-benzimidazol-5-yl)phenylmethanone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Amino-1H-benzimidazol-5-yl)phenylmethanone
CAS 52329-60-9
Analyte or Component Names (2-Amino-1H-benzimidazol-5-yl)phenylmethanone
Shipping Condition Room Temperature
Molecular Formula C14 H11 N3 O
Certifications/Compliance ISO 9001
Synonym (2-Amino-1H-benzimidazol-5-yl)phenylmethanone, 2-Amino-5(6)-benzoylbenzimidazole, Methanone, (2-amino-1H-benzimidazol-5-yl)phenyl- (9CI), (2-Amino-1H-benzimidazol-6-yl)phenylmethanone, 2-Amino-5-benzoyl-1H-benzimidazole, 2-Amino-5-benzoylbenzimidazole, Aminomebendazole, G 1029, R 18986, Mebendazole Related Compound A, Mebendazole Impurity A
InChI Formula InChI=1S/C14H11N3O/c15-14-16-11-7-6-10(8-12(11)17-14)13(18)9-4-2-1-3-5-9/h1-8H,(H3,15,16,17)
SMILES Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3
IUPAC Name (2-amino-1H-benzimidazol-5-yl)-phenylmethanone
Molecular Weight (g/mol) 237.26
Quantity 50 mg
Formula Weight 237.0902 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.