missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Amino-1-propanol, 98%

Product Code. 11322236
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322236 5 g 5g
11332236 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322236 Supplier Thermo Scientific Alfa Aesar Supplier No. L03608.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6168-72-5 | C3H9NO | 75.11 g/mol

DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6168-72-5
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008085
InChI Key BKMMTJMQCTUHRP-UHFFFAOYNA-N
Synonym dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
PubChem CID 5126
IUPAC Name 2-aminopropan-1-ol
SMILES CC(N)CO

Specifications

Melting Point 8°C
Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Refractive Index 1.4495
Quantity 5 g
UN Number UN2735
Beilstein 1209234
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (+/-)-2-Amino-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.