missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Amino-1-propanol-1,1,2,3,3,3-d6-N-t-BOC, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl (1-hydroxypropan-2-yl-1,1,2,3,3,3-d6)carbonate-D6 |
| CAS | 2243764-35-2 |
| Molecular Formula | C8 D6 H11 N O3 |
| Quantity | 0.1 g |
| Synonym | Carbamic acid, N-[1-(hydroxymethyl-d2)ethyl-1,2,2,2-d4]-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl N-[1-(hydroxymethyl-d2)ethyl-1,2,2,2-d4]carbamate (ACI), tert-Butyl N-(1-hydroxypropan-2-yl-1,1,2,3,3,3-d6)carbamate, N-Boc-DL-alaninol-1,1,2,3,3,3-D6, N-tert-Butyloxycarbonyl DL-Alaninol-d6 |
| InChI Formula | InChI=1 S/C8H17NO3/c1-6(5-10)9-7(11)12-8(2,3)4/h6,10 H,5H2,1-4H3,(H,9,11)/i1D3,5D2,6 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NC(=O)OC(C)(C)C)C([2 H])([2 H])O |
| IUPAC Name | tert-butyl N-(1,1,1,2,3,3-hexadeuterio-3-hydroxypropan-2-yl)carbamate |
| Molecular Weight (g/mol) | 181.2625 |
| Formula Weight | 181.1585 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction