missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-phenylethanol, MedChemExpress

Product Code. 16431959
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431959 Supplier Medchem Express Supplier No. HY59132
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Amino-1-phenylethanol is an analogue of noradrenaline.

2-Amino-1-phenylethanol is an analogue of noradrenaline.

Specifications

Chemical Name or Material 2-Amino-1-phenylethanol
Physical Form Solid or Liquid
CAS 7568-93-6
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H11NO
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 100 mg/mL (728.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(CN)C1=CC=CC=C1
Molecular Weight (g/mol) 137.18
Formula Weight 137.18
Percent Purity 95.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.