missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,7-dihydro-7-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, TRC

Product Code. 17902485
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17902485 100 mg 100mg
17903695 25 mg 25mg
17902495 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17902485 Supplier Toronto Research Chemicals Supplier No. TRCA610065100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84222-50-4
Molecular Formula C9 H13 N5 O4
Molecular Weight (g/mol) 255.23
Synonym 2-Amino-7-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-1,7-dihydro-6H-purin-6-one, Ganciclovir Imp. H (EP), N-7 Isomer of Ganciclovir
IUPAC Name 2-amino-7-(1,3-dihydroxypropan-2-yloxymethyl)-1H-purin-6-one
SMILES NC1=Nc2ncn(COC(CO)CO)c2C(=O)N1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)14(3-11-7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)
Formula Weight 255.0968
Chemical Name or Material 2-Amino-7-[(2-hydroxy-1-(hydroxymethyl)ethoxy)methyl]-1,7-dihydro-6H-purin-6-one (N-7 Isomer of Ganciclovir)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.