missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3-propanediol hydrochloride, 98%, Thermo Scientific™

Product Code. 11427106
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427106 0.25 g 0.25g
11417106 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427106 Supplier Thermo Scientific Alfa Aesar Supplier No. B25419.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73708-65-3
Molecular Formula C3H10NO2
Molecular Weight (g/mol) 92.12
MDL Number MFCD00012592
InChI Key KJJPLEZQSCZCKE-UHFFFAOYSA-O
Synonym serinol hydrochloride, 2-aminopropane-1,3-diol hydrochloride, 2-amino-1,3-propanediol hydrochloride, 1,3-propanediol, 2-amino-, hydrochloride, dl-serinol, acmc-1bl7f, 2-amino-1,3-propanediol hyrochloride
PubChem CID 2734815
IUPAC Name 2-aminopropane-1,3-diol;hydrochloride
SMILES [NH3+]C(CO)CO

Specifications

Melting Point 104°C to 106°C
Quantity 0.25 g
Sensitivity Hygroscopic
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Amino-1,3-propanediol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.