missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.
Solubility
Soluble in water and alcohol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.
Solubility
Soluble in water and alcohol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Melting Point | 52°C to 56°C |
Boiling Point | 128°C (0.8 mmHg) |
Quantity | 5 g |
UN Number | UN3259 |
Beilstein | 635683 |
Sensitivity | Air sensitive; Hygroscopic |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 91.11 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-1,3-propanediol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'