missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3-propanediol, 98%

Product Code. 11358155
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358155 1 g 1g
11368155 5 g 5g
11378155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358155 Supplier Thermo Scientific Alfa Aesar Supplier No. L08426.03
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 534-03-2 | C3H9NO2 | 91.11 g/mol

2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3-propanediol is used as an intermediate in several chemical applications and also useful for serinol and some of its derivatives.

Solubility
Soluble in water and alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-03-2
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00051487
InChI Key KJJPLEZQSCZCKE-UHFFFAOYSA-N
Synonym 2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
PubChem CID 68294
IUPAC Name 2-aminopropane-1,3-diol
SMILES NC(CO)CO

Specifications

Melting Point 52°C to 56°C
Boiling Point 128°C (0.8 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 635683
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material 2-Amino-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.