missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3,4-thiadiazole, 98+%

Product Code. 11384514
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384514 25 g 25g
11374514 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384514 Supplier Thermo Scientific Alfa Aesar Supplier No. L01244.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4005-51-0 | C2H3N3S | 101.127 g/mol

2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

Solubility
It is soluble in water, hot methanol- very faint turbidity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4005-51-0
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.127
MDL Number MFCD00003107
InChI Key QUKGLNCXGVWCJX-UHFFFAOYSA-N
Synonym 2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
PubChem CID 19909
IUPAC Name 1,3,4-thiadiazol-2-amine
SMILES C1=NN=C(S1)N

Specifications

Melting Point 188°C to 194°C
Quantity 25 g
UN Number UN2811
Beilstein 107135
Solubility Information It is soluble in water,hot methanol- very faint turbidity.
Formula Weight 101.13
Percent Purity ≥98%
Chemical Name or Material 2-Amino-1,3,4-thiadiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.