missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3,4-thiadiazole, 97%, Thermo Scientific™

Product Code. 10219743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219743 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219743 Supplier Thermo Scientific Acros Supplier No. 268160010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4005-51-0
MDL Number MFCD00003107
InChI Key QUKGLNCXGVWCJX-UHFFFAOYSA-N
Synonym 2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
PubChem CID 19909
IUPAC Name 1,3,4-thiadiazol-2-amine
SMILES C1=NN=C(S1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 189.0°C to 192.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C2H3N3S
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents,alcohol
Molecular Weight (g/mol) 101.13
Formula Weight 101.13
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Amino-1, 3, 4-thiadiazole, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-1,3,4-thiadiazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H371-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.