missing translation for 'onlineSavingsMsg'
Learn More

2-(Alpha-D-Mannopyranosyl)-L-tryptophan, TRC

Product Code. 30004851
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004851 1 mg 1mg
30136381 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004851 Supplier Toronto Research Chemicals Supplier No. TRCM1664501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180509-18-6
Molecular Formula C17H22N2O7
Molecular Weight (g/mol) 366.37
Synonym C2-α-D-Mannopyranosyltryptophan, 2-α-D-Mannopyranosyl-L-tryptophan, 2-α-D-Mannopyranosyl-L-tryptophan
IUPAC Name (S)-2-amino-3-(2-((2R,3S,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=C([C@@H]2[C@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)NC3=C1C=CC=C3)C(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H22N2O7/c18-9(17(24)25)5-8-7-3-1-2-4-10(7)19-12(8)16-15(23)14(22)13(21)11(6-20)26-16/h1-4,9,11,13-16,19-23H,5-6,18H2,(H,24,25)/t9-,11+,13+,14-,15-,16+/m0/s1
Formula Weight 366.14
Percent Purity >95%
Chemical Name or Material 2-(alpha-D-Mannopyranosyl)-L-tryptophan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.