missing translation for 'onlineSavingsMsg'
Learn More

2-Allyloxybenzaldehyde, 97%

Product Code. 11456157
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456157 25 g 25g
11446157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456157 Supplier Thermo Scientific Alfa Aesar Supplier No. B25119.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 28752-82-1 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 28752-82-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00014130
InChI Key BXCJDECTRRMSCV-UHFFFAOYSA-N
Synonym 2-allyloxy benzaldehyde, 2-allyloxybenzaldehyde, o-allyloxy benzaldehyde, o-allyloxybenzaldehyde, benzaldehyde, 2-2-propenyloxy, benzaldehyde, o-allyloxy, 2-prop-2-en-1-yloxy benzaldehyde, 2-prop-2-enyloxybenzaldehyde, allylsalicylaldehyde, o-allylsalicylaldehyde
PubChem CID 101335
IUPAC Name 2-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=CC=C1C=O

Tekniske data

Density 1.079
Boiling Point 129°C to 132°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.55
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 2-Allyloxybenzaldehyde

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.