missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-N-Boc-L-glycine dicyclohexylamine salt, 95%, Thermo Scientific™

Product Code. 15469545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469545 5 g 5g
15459545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469545 Supplier Thermo Scientific Alfa Aesar Supplier No. H52062.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143979-15-1
Molecular Formula C22H40N2O4
Molecular Weight (g/mol) 396.572
MDL Number MFCD01321013
InChI Key VMCGMPITVQIMGK-ZLTKDMPESA-N
Synonym dicyclohexylamine s-2-tert-butoxycarbonyl amino pent-4-enoate, boc-l-2-allylglycine dicyclohexylamine salt, boc-l-allylglycine.dcha, boc-l-allylglycine-dcha, boc-l-allylglycine dicyclohexylaminoium salt, boc-d-allylglycine-dcha, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid; dicha, boc-l-allylglycine dcha, boc-algly-oh dcha, boc-l-allyglycine?cha
PubChem CID 2755982
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Quantity 5 g
Formula Weight 396.57
Percent Purity 95%
Chemical Name or Material 2-Allyl-N-Boc-L-glycine dicyclohexylamine salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.