missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-3-hydroxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 10362333
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10362333 10 g 10g
10558283 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10362333 Supplier Thermo Scientific Maybridge Supplier No. BTB09979EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 79950-42-8
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00100646
InChI Key QVHRAGBOMUXWRI-UHFFFAOYSA-N
Synonym 2-allyl-3-hydroxybenzaldehyde, 3-hydroxy-2-prop-2-en-1-yl benzaldehyde, 2-allyl-3hydroxybenzaldehyde, 2-allyl-3-hydroxy-benzaldehyde, 3-hydroxy-2-allyl benzaldehyde, 3-oxidanyl-2-prop-2-enyl-benzaldehyde, 3-hydroxy-2-prop-2-en-1-ylbenzaldehyde, benzaldehyde, 3-hydroxy-2-2-propenyl, benzaldehyde,3-hydroxy-2-2-propen-1-yl, 2-allyl-3-hydroxybenzaldehyde, containing 200ppm mehq
PubChem CID 2775156
IUPAC Name 3-hydroxy-2-prop-2-enylbenzaldehyde
SMILES OC1=CC=CC(C=O)=C1CC=C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 2-Allyl-3-hydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.