Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Product Code. 15214283
Click to view available options
Connection Type:
M24 x 1.5 F to M24 x 1.5 F
M24 x 1.5 F to M30 x 1.5 F
M24 x 1.5 F to NPT 1 in. F
M24 x 1.5 F to NPT 1/2 in. M
M24 x 1.5 F to NPT 1/4 in. F
M24 x 1.5 F to NPT 1/4 in. M
M24 x 1.5 F to NPT 3/4 in. F
M24 x 1.5 F to NPT 3/4 in. M
M24 x 1.5 F to NPT 3/8 in. F
M24 x 1.5 Female to 1 in. Tube
M24 x 1.5 Female to 1/4 in. Tube
M24 x 1.5 Female to 3/8 in. Tube
M24 x 1.5 Female to M30 x 1.5 Male
M24 x 1.5 Female to NPT 1 in. Male
M24 x 1.5 Male to M16 x 1.5 Female
M24 x 1.5 Male to M30 x 1.5 Female
M30 x 1.5 F to M30 x 1.5 F
M30 x 1.5 F to M38 x 1.5 F
M30 x 1.5 F to NPT 1 in. F
M30 x 1.5 F to NPT 3/4 in. F
M30 x 1.5 F to NPT 3/4 in. M
M30 x 1.5 Female to 1 in. Tube
M30 x 1.5 Female to M38 x 1.5 Male
M30 x 1.5 Female to NPT 1 in. Male
M30 x 1.5 Male to M38 x 1.5 Female
M30 x 1.5 Male to Male
M38 x 1.5 F to M38 x 1.5 F
M38 x 1.5 F to NPT 1 in. F
M38 x 1.5 F to NPT1 1/4 in. F
M38 x 1.5 F to NPT1 1/4 in. M
M38 x 1.5 Female to 1 in. Tube
M38 x 1.5 Female to NPT 1 in. Male
M38 x 1.5 Male to Male
NPT 3/4 in. Female to M16 x 1 Male
Unit Size:
Pack of 2
Product Code. 15214283
Brand: Julabo™ 8890083
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
Type 2-Adapters
Connection Type M30 x 1.5 F to NPT 3/4 in. M