missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanol, 98%

Product Code. 10408740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10408740 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10408740 Supplier Thermo Scientific Acros Supplier No. 167310050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 700-57-2 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 700-57-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 258.0°C to 265.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H16O
MDL Number MFCD00074744
Quantity 5 g
Synonym 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol, diethyl ether, methylene, chloride and acetone
InChI Key FOWDOWQYRZXQDP-UHFFFAOYSA-N
SMILES C1C2CC3CC1CC(C2)C3O
IUPAC Name adamantan-2-ol
Molecular Weight (g/mol) 152.24
PubChem CID 64149
Formula Weight 152.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.