missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanol, 98%

Product Code. 11433464
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433464 25 g 25g
11423464 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433464 Supplier Thermo Scientific Alfa Aesar Supplier No. A10697.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 700-57-2 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-57-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00074744
InChI Key FOWDOWQYRZXQDP-UHFFFAOYSA-N
Synonym 2-adamantanol, 2-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-2-ol, tricyclo 3.3.1.1∼3,7∼ decan-2-ol, 1r,3r,5r,7r-adamantan-2-ol, 1-adamantanol;, 2alpha-adamantanol, pubchem8753
PubChem CID 64149
IUPAC Name adamantan-2-ol
SMILES C1C2CC3CC1CC(C2)C3O

Specifications

Melting Point 299°C to 303°C (sealed tube)
Quantity 25 g
Beilstein 1903957
Formula Weight 152.24
Percent Purity 98%
Chemical Name or Material 2-Adamantanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.