missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Adamantanamine Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-Adamantanamine Hydrochloride |
| CAS | 10523-68-9 |
| Molecular Formula | C10H17N . HCl |
| Quantity | 10 mg |
| Synonym | 2-Adamantaneammonium Chloride, 2-Adamantylamine Hydrochloride, 2-Aminoadamantane Hydrochloride |
| InChI Formula | InChI=1S/C10H17N.ClH/c11-10-8-2-6-1-7(4-8)5-9(10)3-6;/h6-10H,1-5,11H2;1H |
| SMILES | Cl.NC1C2CC3CC(CC1C3)C2 |
| IUPAC Name | adamantan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 187.71 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction