missing translation for 'onlineSavingsMsg'
Learn More

2-Acrylamido-2-methylpropanesulfonic acid, 97%

Product Code. 10517172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10517172 5 g Glass bottle 5g
10330134 50 g Glass bottle 50g
10570934 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10517172 Supplier Thermo Scientific Acros Supplier No. 400290050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15214-89-8 | C7H13NO4S | 207.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15214-89-8
Molecular Formula C7H13NO4S
Molecular Weight (g/mol) 207.24
MDL Number MFCD00007522
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 195.0°C
Color White
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH2CHCONHC(CH3)2CH2SO3H
Quantity 5 g
Solubility Information Solubility in water: 1500g/L (20°C)
Formula Weight 207.25
Percent Purity 97%
Physical Form Powder, Chunks or Chips
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Acrylamido-2-methylpropanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.