missing translation for 'onlineSavingsMsg'
Learn More

2-Acrylamide-2-methylpropanesulfonic Acid, TRC

Product Code. 30315637
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315637 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315637 Supplier Toronto Research Chemicals Supplier No. TRCA19132825MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Acrylamido-2-methylpropanesulphonic acid
CAS 15214-89-8
Molecular Formula C7 H13 N O4 S
Quantity 25 mg
Synonym 1-Propanesulfonic acid, 2-acrylamido-2-methyl- (8CI), 1-Propanesulfonic acid, 2-methyl-2-[(1-oxo-2-propenyl)amino]- (9CI), 2-Methyl-2-[(1-oxo-2-propen-1-yl)amino]-1-propanesulfonic acid, 2-Acrylamido-2,2-dimethylethanesulfonic acid, 2-Acrylamido-2-methyl-1-propanesulfonic acid, 2-Acrylamido-2-methylpropanesulfonic acid, 2-Acrylamido-2-methylpropylsulfonic acid, 2-Acryloylamido-2-methylpropanesulfonic acid, 2-Acryloylamino-2-methyl-1-propanesulfonic acid, 2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid, AMPS, AMPS (sulfonic acid), ATBS, Acrylamide tert-butylsulfonic acid, CG 810S-P, Lubrizol 2402, Lubrizol 2404, Lubrizol AMPS, TBAS-Q, tert-Butylacrylamidosulfonic acid
InChI Formula InChI=1S/C7H13NO4S/c1-4-6(9)8-7(2,3)5-13(10,11)12/h4H,1,5H2,2-3H3,(H,8,9)(H,10,11,12)
SMILES CC(C)(CS(=O)(=O)O)NC(=O)C=C
IUPAC Name 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
Molecular Weight (g/mol) 207.247
Formula Weight 207.0565
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.