missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 99%

Product Code. 11464364
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464364 2500 g 2500g
11444364 100 g 100g
11454364 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464364 Supplier Thermo Scientific Alfa Aesar Supplier No. A13062.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 88-15-3 | C6H6OS | 126.173 g/mol

2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1

Specifications

Melting Point 10°C to 12°C
Density 1.168
Boiling Point 213°C to 214°C
Flash Point 91°C (195°F)
Odor Slightly Sweetish
Refractive Index 1.566
Quantity 2500 g
UN Number UN2810
Beilstein 107910
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 126.18
Percent Purity 99%
Chemical Name or Material 2-Acetylthiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.