missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 98%

Product Code. 10667893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667893 500 g Glass bottle 500g
10467280 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10667893 Supplier Thermo Scientific Acros Supplier No. 102725000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-15-3 | C6H6OS | 126.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 10°C to 11°C
Color Brown to Yellow
Density 1.1600g/mL
Boiling Point 214°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C4H3S)COCH3
Refractive Index 1.5640 to 1.566
Quantity 500 g
Beilstein 17, 287
Specific Gravity 1.16
Solubility Information Solubility in water: 14g/L (30°C). Other solubilities: soluble in ethanol,benzene,chloroform,benzene
Formula Weight 126.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Acetylthiophene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.