missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiazole, 99%

Product Code. 11403624
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403624 1 g 1g
11413624 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403624 Supplier Thermo Scientific Alfa Aesar Supplier No. A11266.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24295-03-2 | C5H5NOS | 127.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24295-03-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00005324
InChI Key MOMFXATYAINJML-UHFFFAOYSA-N
Synonym 2-acetylthiazole, 1-1,3-thiazol-2-yl ethan-1-one, ethanone, 1-2-thiazolyl, 1-1,3-thiazol-2-yl ethanone, 2-acetyl thiazole, 1-2-thiazolyl ethanone, ketone, methyl 2-thiazolyl, 2-acetylthiazol, 2-acetyl-1,3-thiazole, 1-thiazol-2-yl-ethanone
PubChem CID 520108
IUPAC Name 1-(1,3-thiazol-2-yl)ethanone
SMILES CC(=O)C1=NC=CS1

Specifications

Density 1.227
Boiling Point 89°C to 91°C (12 mmHg)
Flash Point 78°C (172°F)
Odor Characteristic
Refractive Index 1.548
Quantity 1 g
Beilstein 109803
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material 2-Acetylthiazole
compliance-icons
Product Identifier
  • 2-Acetylthiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Acetylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.