missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetyloxy)-3-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. p-2696341
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10712693

Brand: Thermo Scientific Maybridge SB01734DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4386-39-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00085683
InChI Key XRBMKGUDDJPAMH-UHFFFAOYSA-N
Synonym 2-acetyloxy-3-methylbenzoic acid, 2-acetoxy-3-methylbenzoic acid, 2-acetoxy-m-toluic acid, unii-2g83e641hz, 2-acetoxy-3-methyl-benzoic acid, 3-methylacetylsalicylic acid, benzoic acid,2-acetyloxy-3-methyl, 3-methyl-2-acetoxybenzoic acid, cresopirine, cresopyrine
PubChem CID 78094
IUPAC Name 2-acetyloxy-3-methylbenzoic acid
SMILES CC(=O)OC1=C(C)C=CC=C1C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 2-(Acetyloxy)-3-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.