missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetyloxy)-3-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10269924
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10269924 5 g 5g
10712693 1 g 1g
10633223 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269924 Supplier Thermo Scientific Maybridge Supplier No. SB01734DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4386-39-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00085683
InChI Key XRBMKGUDDJPAMH-UHFFFAOYSA-N
Synonym 2-acetyloxy-3-methylbenzoic acid, 2-acetoxy-3-methylbenzoic acid, 2-acetoxy-m-toluic acid, unii-2g83e641hz, 2-acetoxy-3-methyl-benzoic acid, 3-methylacetylsalicylic acid, benzoic acid,2-acetyloxy-3-methyl, 3-methyl-2-acetoxybenzoic acid, cresopirine, cresopyrine
PubChem CID 78094
IUPAC Name 2-acetyloxy-3-methylbenzoic acid
SMILES CC(=O)OC1=C(C)C=CC=C1C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 2-(Acetyloxy)-3-methylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.