missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylfuran, TRC

Product Code. 30134869
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134869 1 g 1g
18062530 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134869 Supplier Toronto Research Chemicals Supplier No. TRCA1762001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6 H6 O2
Molecular Weight (g/mol) 110.11
Synonym Ketone, 2-furyl methyl (8CI), 1-(2-Furanyl)ethanone, 1-(2-Furyl)ethanone, 2-Acetylfuran, 2-Furyl methyl ketone, 2-Furylethanone, Methyl 2-furyl ketone, NSC 4665, NSC 49133, α-Acetylfuran
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)c1occc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6O2/c1-5(7)6-3-2-4-8-6/h2-4H,1H3
Formula Weight 110.0368
Chemical Name or Material 2-Acetylfuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.