missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylfuran, 99%

Product Code. 10192410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192410 250 g Glass bottle 250g
10796121 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192410 Supplier Thermo Scientific Acros Supplier No. 102552500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-62-7 | C6H6O2 | 110.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-62-7
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00003242
InChI Key IEMMBWWQXVXBEU-UHFFFAOYSA-N
Synonym 2-acetylfuran, acetylfuran, 1-furan-2-yl ethanone, 2-furyl methyl ketone, methyl 2-furyl ketone, 1-2-furyl ethanone, 2-furylethanone, 2-acetyl furan, ketone, 2-furyl methyl, ethanone, 1-2-furanyl
PubChem CID 14505
ChEBI CHEBI:59983
IUPAC Name 1-(furan-2-yl)ethanone
SMILES CC(=O)C1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Color Yellow to Brown
Density 1.0980g/mL
Boiling Point 67.0°C (10.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. 2,5-Diacetyl furan (GC)
Packaging Glass bottle
Refractive Index 1.5060 to 1.5080
Quantity 250 g
Beilstein 17, 286
Specific Gravity 1.098
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 110.11
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Acetylfuran, 99%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.