missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbutyrolactone, Mikromol™

Product Code. 17984046
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17984046 Supplier Mikromol Supplier No. MM0491.250100
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Acetylbutyrolactone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Acetylbutyrolactone
CAS 517-23-7
Analyte or Component Names 2-Acetylbutyrolactone
Shipping Condition +4°C
Molecular Formula C6H8O3
Certifications/Compliance ISO 9001
Synonym 3-Acetyldihydro-2(3H)-furanone, 3-Acetyldihydrofuran-2-one, alpha-Acetyl-gamma-butyrolactone, alpha-Acetyl-gamma-hydroxybutyric Acid gamma-Lactone, 2-Oxo-3-acetyltetrahydrofuran, NSC 2019, alpha-(2-Hydroxyethyl)acetoacetic acid gamma-lactone
SMILES CC(=O)C1CCOC1=O
Molecular Weight (g/mol) 128.13
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.