missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzoic acid, 98+%

Product Code. 11409552
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409552 250 g 250g
11489542 10 g 10g
11499542 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409552 Supplier Thermo Scientific Alfa Aesar Supplier No. A10216.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-56-0 | C9H8O3 | 164.16 g/mol

2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-56-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002475
InChI Key QDAWXRKTSATEOP-UHFFFAOYSA-N
Synonym o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
PubChem CID 68474
IUPAC Name 2-acetylbenzoic acid
SMILES CC(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 115°C to 118°C
Odor Odorless
Quantity 250 g
Beilstein 1210557
Solubility Information Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
Formula Weight 164.16
Percent Purity ≥98%
Chemical Name or Material 2-Acetylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.